CH6371383324 Chem Impex International, Inc.

2-Methyl-5,6-dihydro-4H-pyrrolo[3,4-d]thiazole hydrochloride

  • MDL number MFCD11506339

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-methyl-5-6-dihydro-4-i-h-i-pyrrolo-3-4-i-delta-i-thiazole-hydrochloride/9816?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-methyl-5-6-dihydro-4-i-h-i-pyrrolo-3-4-i-delta-i-thiazole-hydrochloride/9816?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart