CH6371383329 Chem Impex International, Inc.

2-m-Tolyl-thiazole-4-carboxylic acid ethyl ester

  • MDL number MFCD06738345
  • Cas Number : 132089-33-9
  • Molecular Weight : 247.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8 °C
Physical Form Yellow solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-i-m-i-tolyl-thiazole-4-carboxylic-acid-ethyl-ester/9829?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-i-m-i-tolyl-thiazole-4-carboxylic-acid-ethyl-ester/9829?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart