CH6371383331 Chem Impex International, Inc.

2-Nitro-benzoic acid tert-butyl ester

  • MDL number MFCD06291302
  • Cas Number : 55666-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form Orange liquid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-nitro-benzoic-acid-i-tert-i-butyl-ester/9831?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-nitro-benzoic-acid-i-tert-i-butyl-ester/9831?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart