CH6371383332 Chem Impex International, Inc.

2-Nitrophenethylamine

  • MDL number MFCD06212598
  • Cas Number : 33100-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form Brown liquid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-nitrophenethylamine/9832?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-nitrophenethylamine/9832?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart