CH6371383347 Chem Impex International, Inc.

3-(1H-Imidazol-4-yl)benzylamine

  • MDL number MFCD06738849
  • Cas Number : 885281-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8°C
Physical Form Off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-1h-imidazol-4-yl-benzylamine/9878?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-1h-imidazol-4-yl-benzylamine/9878?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart