CH6371383362 Chem Impex International, Inc.

(S)-3-Amino-2(benzylamino)propan-1-ol

  • MDL number MFCD06658298
  • Cas Number : 150482-72-7
  • Molecular Weight : 180.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Physical Form Oil
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-2-benzylamino-propan-1-ol/9923?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-2-benzylamino-propan-1-ol/9923?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart