CH6371383365 Chem Impex International, Inc.

3,5-Dinitro-benzoic acid tert-butyl ester

  • MDL number MFCD00432164
  • Cas Number : 5342-97-2
  • Molecular Weight : 268.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8 °C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-5-dinitro-benzoic-acid-i-tert-i-butyl-ester/9932?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-5-dinitro-benzoic-acid-i-tert-i-butyl-ester/9932?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart