CH6371383376 Chem Impex International, Inc.

3-Bromo-4-oxo-piperidine-1-carboxylic acid ethyl ester

  • MDL number MFCD06796218
  • Cas Number : 95629-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Physical Form Yellowish liquid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-bromo-4-oxo-piperidine-1-carboxylic-acid-ethyl-ester/9952?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-bromo-4-oxo-piperidine-1-carboxylic-acid-ethyl-ester/9952?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart