CH6371383385 Chem Impex International, Inc.

3-Chloro-2-iodophenylamine

  • MDL number MFCD06738966
  • Cas Number : 70237-25-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8
Physical Form Dark-brown liquid to crystalline
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-chloro-2-iodophenylamine/9963?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-chloro-2-iodophenylamine/9963?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart