CH6371383393 Chem Impex International, Inc.

3-Fluorobenzamidine hydrochloride

  • MDL number MFCD04114433

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Fluorobenzenecarboximidamide hydrochloride
Purity 97
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-fluorobenzamidine-hydrochloride/9975?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-fluorobenzamidine-hydrochloride/9975?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart