CH6371383408 Chem Impex International, Inc.

3-Phenyl-pyridin-2-ylamine

  • MDL number MFCD04114119
  • Cas Number : 87109-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Amino-3-phenylpyridine
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-phenyl-pyridin-2-ylamine/10010?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-phenyl-pyridin-2-ylamine/10010?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart