CH6371383410 Chem Impex International, Inc.

4-(2'-Methyl-4'-nitrophenoxy) piperidine hydrochloride

  • MDL number MFCD06658484
  • Cas Number : 138227-67-5
  • Molecular Weight : 272.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form Brown solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-2-methyl-4-nitrophenoxy-piperidine-hydrochloride/10019?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-2-methyl-4-nitrophenoxy-piperidine-hydrochloride/10019?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart