CH6371383419 Chem Impex International, Inc.

4-Amino-3-bromo-5-iodo-benzoic acid ethyl ester

  • MDL number MFCD06658437
  • Cas Number : 437707-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ethyl 4-amino-3-bromo-5-iodobenzoate
Purity 0.95
Storage temp. Store at 0-8
Physical Form White or brown solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-amino-3-bromo-5-iodo-benzoic-acid-ethyl-ester/10042?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-amino-3-bromo-5-iodo-benzoic-acid-ethyl-ester/10042?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart