CH6371383423 Chem Impex International, Inc.

4-Benzyl-2-((pyrrolidin-1-yl)methyl) morpholine

  • MDL number MFCD06658347
  • Cas Number : 635699-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form Reddish liquid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-benzyl-2-pyrrolidin-1-yl-methyl-morpholine/10048?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-benzyl-2-pyrrolidin-1-yl-methyl-morpholine/10048?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart