CH6371383426 Chem Impex International, Inc.

(3-Bromo-pyridin-4-yl)carbamic acid tert-butyl ester

  • MDL number MFCD01860257
  • Cas Number : 257937-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl 3-bromopyridin-4-ylcarbamate
Purity 95
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-bromo-pyridin-4-yl-carbamic-acid-i-tert-i-butyl-ester/10052?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-bromo-pyridin-4-yl-carbamic-acid-i-tert-i-butyl-ester/10052?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart