CH6371383443 Chem Impex International, Inc.

4-Fluoro-3-nitro-benzylamine

  • MDL number MFCD06212995
  • Cas Number : 771581-73-8
  • Molecular Weight : 170.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Physical Form Yellow solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-fluoro-3-nitro-benzylamine/10071?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-fluoro-3-nitro-benzylamine/10071?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart