CH6371383449 Chem Impex International, Inc.

3-Cyano-4-iminopiperidine

  • MDL number MFCD06658412
  • Cas Number : 20373-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0 - 8 °C
Physical Form Light yellow solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-cyano-4-iminopiperidine/10079?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-cyano-4-iminopiperidine/10079?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart