CH6371383468 Chem Impex International, Inc.

4-Phenyl-azepane

  • MDL number MFCD08060059
  • Cas Number : 73252-01-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Physical Form Colorless liquid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-phenyl-azepane/10099?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-phenyl-azepane/10099?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart