CH6371383485 Chem Impex International, Inc.

5-(4-Cyanobenzoylamino)-2-ethoxy-benzenesulfonyl chloride

  • MDL number MFCD03424962
  • Cas Number : 680618-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form Dark brown solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-4-cyanobenzoylamino-2-ethoxy-benzenesulfonyl-chloride/10127?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-4-cyanobenzoylamino-2-ethoxy-benzenesulfonyl-chloride/10127?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart