CH6371383495 Chem Impex International, Inc.

5-Amino-2-(phenylthio)benzoic acid

  • MDL number MFCD06658325
  • Cas Number : 63185-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8 °C
Physical Form Light brown solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-amino-2-phenylthio-benzoic-acid/10143?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-amino-2-phenylthio-benzoic-acid/10143?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart