CH6371383538 Chem Impex International, Inc.

5-Methoxy-1H-indole-3-carboxylic acid methyl ester

  • MDL number MFCD06203662
  • Cas Number : 172595-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl 5-methoxy-1H-indole-3-carboxylate
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-methoxy-1-i-h-i-indole-3-carboxylic-acid-methyl-ester/10196?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-methoxy-1-i-h-i-indole-3-carboxylic-acid-methyl-ester/10196?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart