CH6371383542 Chem Impex International, Inc.

5-Methyl-1,3-dihydro-2H-indol-2-one

  • MDL number MFCD02379906
  • Cas Number : 3484-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Methyl-1,3-dihydro-indol-2-one
Purity 95
Storage temp. Store at 0-8
Physical Form Brown crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-methyl-1-3-dihydro-2-i-h-i-indol-2-one/10200?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-methyl-1-3-dihydro-2-i-h-i-indol-2-one/10200?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart