CH6371383551 Chem Impex International, Inc.

5-Nitro-1H-indazole-7-carboxylic acid methyl ester

  • MDL number MFCD03990536
  • Cas Number : 632291-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8°C
Physical Form Yellow to beige solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-nitro-1-i-h-i-indazole-7-carboxylic-acid-methyl-ester/10212?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-nitro-1-i-h-i-indazole-7-carboxylic-acid-methyl-ester/10212?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart