CH6371383607 Chem Impex International, Inc.

6-Methoxy-1,2,3,4-tetrahydronaphthalen-2-ylamine

  • MDL number MFCD04114915
  • Cas Number : 81861-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8 °C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/6-methoxy-1-2-3-4-tetrahydronaphthalen-2-ylamine/10289?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/6-methoxy-1-2-3-4-tetrahydronaphthalen-2-ylamine/10289?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart