CH6371383618 Chem Impex International, Inc.

6-Phenyl-piperidine-2-carboxylic acid

  • MDL number MFCD06410516
  • Cas Number : 791559-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8°C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/6-phenyl-piperidine-2-carboxylic-acid/10310?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/6-phenyl-piperidine-2-carboxylic-acid/10310?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart