CH6371383738 Chem Impex International, Inc.

Methyl 2-bromomethyl-6-chlorobenzoate

  • MDL number MFCD04114325
  • Cas Number : 482578-63-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8
Physical Form Oil
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/methyl-2-bromomethyl-6-chlorobenzoate/10507?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/methyl-2-bromomethyl-6-chlorobenzoate/10507?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart