CH6371383767 Chem Impex International, Inc.

Oxazole-4-carbothioic acid amide

  • MDL number MFCD06738920
  • Cas Number : 118802-31-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form Pale yellow solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/oxazole-4-carbothioic-acid-amide/10544?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/oxazole-4-carbothioic-acid-amide/10544?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart