CH6371383772 Chem Impex International, Inc.

3-Methylbenzoyl cyanide

  • MDL number MFCD06658475
  • Cas Number : 5955-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-methylbenzoyl-cyanide/10549?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-methylbenzoyl-cyanide/10549?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart