CH6371383773 Chem Impex International, Inc.

2-Methylbenzoyl cyanide

  • MDL number MFCD06658474
  • Cas Number : 5955-73-7
  • Molecular Weight : 145.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form Yellow liquid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-methylbenzoyl-cyanide/10550?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-methylbenzoyl-cyanide/10550?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart