CH6371383774 Chem Impex International, Inc.

4-Methylbenzoyl cyanide

  • MDL number MFCD00182410
  • Cas Number : 14271-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8°C
Physical Form Yellow solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-methylbenzoyl-cyanide/10551?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-methylbenzoyl-cyanide/10551?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart