CH6371383793 Chem Impex International, Inc.

Quinoline-4-carbothioic acid amide

  • MDL number MFCD02179220
  • Cas Number : 74585-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Physical Form Yellow solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/quinoline-4-carbothioic-acid-amide/10577?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/quinoline-4-carbothioic-acid-amide/10577?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart