CH6371383796 Chem Impex International, Inc.

Quinoline-8-carbothioic acid amide

  • MDL number MFCD06738915
  • Cas Number : 62216-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Physical Form Yellow solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/quinoline-8-carbothioic-acid-amide/10581?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/quinoline-8-carbothioic-acid-amide/10581?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart