CH6371383845 Chem Impex International, Inc.

a-Tosyl-(2-bromobenzyl)isocyanide

  • MDL number MFCD04114784
  • Cas Number : 936548-16-2
  • Molecular Weight : 350.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8°C
Physical Form Grey solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/a-tosyl-2-bromobenzyl-isocyanide/10733?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/a-tosyl-2-bromobenzyl-isocyanide/10733?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart