CH6371383956 Chem Impex International, Inc.

2,4-Dichloro-5H-pyrrolo[3,2-d]pyrimidine

  • MDL number MFCD08059281
  • Cas Number : 63200-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at RT
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-4-dichloro-5h-pyrrolo-3-2-d-pyrimidine/11008?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-4-dichloro-5h-pyrrolo-3-2-d-pyrimidine/11008?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart