CH6371383987 Chem Impex International, Inc.

2-Chloro-4-fluoro-benzamidine hydrochloride

  • MDL number MFCD04114447
  • Cas Number : 582306-90-9
  • Molecular Weight : 209.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0 - 8 °C
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-chloro-4-fluoro-benzamidine-hydrochloride/11075?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-chloro-4-fluoro-benzamidine-hydrochloride/11075?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart