CH6371383999 Chem Impex International, Inc.

2-Methyl-5-nitro-benzoic acid methyl ester

  • MDL number MFCD00230865
  • Cas Number : 77324-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Methyl-5-nitro methylbenzoate
Purity 99
Storage temp. Store at 0-8
Physical Form Yellow solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-methyl-5-nitro-benzoic-acid-methyl-ester/11103?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-methyl-5-nitro-benzoic-acid-methyl-ester/11103?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart