CH6371384113 Chem Impex International, Inc.

4-Dimethylamino-benzamidine hydrochloride

  • MDL number MFCD04114415
  • Cas Number : 55978-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Physical Form Off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-dimethylamino-benzamidine-hydrochloride/11350?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-dimethylamino-benzamidine-hydrochloride/11350?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart