CH6371384122 Chem Impex International, Inc.

4-Methoxy-2-methyl-benzoic acid methyl ester

  • MDL number MFCD06203918
  • Cas Number : 35598-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Physical Form Colorless to pale yellow liquid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Boiling point 141-145� C
Product Link http://www.chemimpex.com/product/productinfo/4-methoxy-2-methyl-benzoic-acid-methyl-ester/11373?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-methoxy-2-methyl-benzoic-acid-methyl-ester/11373?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart