CH6371384133 Chem Impex International, Inc.

5-(4-Methylsulfanyl-phenyl)-1-phenyl-1H-pyrazole-3-carboxylic acid methyl ester

  • MDL number MFCD04115083
  • Cas Number : 640727-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103-110 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White amorphous powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-4-methylsulfanyl-phenyl-1-phenyl-1h-pyrazole-3-carboxylic-acid-methyl-ester/11404?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-4-methylsulfanyl-phenyl-1-phenyl-1h-pyrazole-3-carboxylic-acid-methyl-ester/11404?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart