CH6371384138 Chem Impex International, Inc.

5,6,7,8-Tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine oxalate

  • MDL number MFCD07371490
  • Cas Number : 59624-08-7
  • Molecular Weight : 214.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8 °C
Physical Form Off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-6-7-8-tetrahydro-1-2-4-triazolo-4-3-a-pyrazine-oxalate/11414?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-6-7-8-tetrahydro-1-2-4-triazolo-4-3-a-pyrazine-oxalate/11414?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart