CH6371384259 Chem Impex International, Inc.

7-Hydroxy-3,4-dihydro-2H-isoquinolin-1-one

  • MDL number MFCD04114872
  • Cas Number : 22246-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Physical Form Off-white crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/7-hydroxy-3-4-dihydro-2h-isoquinolin-1-one/11655?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/7-hydroxy-3-4-dihydro-2h-isoquinolin-1-one/11655?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart