CH6371384285 Chem Impex International, Inc.

5-Aminomethyl-3-isopropylisoxazole

  • MDL number MFCD07643181
  • Cas Number : 543713-30-0
  • Molecular Weight : 140.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form Liquid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-aminomethyl-3-isopropylisoxazole/11715?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-aminomethyl-3-isopropylisoxazole/11715?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart