CH6371384327 Chem Impex International, Inc.

Methyl 5-(4-methylsulfanylphenyl)-1-(4-trifluoromethyl-phenyl)-1H-pyrazole-3-carboxylate

  • MDL number MFCD04973408
  • Cas Number : 641639-55-6
  • Molecular Weight : 392.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8 °C
Physical Form Pale yellow solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/methyl-5-4-methylsulfanylphenyl-1-4-trifluoromethyl-phenyl-1h-pyrazole-3-carboxylate/11835?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/methyl-5-4-methylsulfanylphenyl-1-4-trifluoromethyl-phenyl-1h-pyrazole-3-carboxylate/11835?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart