CH6371384330 Chem Impex International, Inc.

Methyl 5-iodo-1H-indazole-3-carboxylate

  • MDL number MFCD07371578
  • Cas Number : 1079-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8°C
Physical Form Off-white to grey solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/methyl-5-iodo-1h-indazole-3-carboxylate/11844?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/methyl-5-iodo-1h-indazole-3-carboxylate/11844?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart