CH6371384360 Chem Impex International, Inc.

N-O-Tolyl-guanidine

  • MDL number MFCD04114636
  • Cas Number : 37557-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8 °C
Physical Form Off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/n-o-tolyl-guanidine/11902?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/n-o-tolyl-guanidine/11902?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart