CH6371384362 Chem Impex International, Inc.

N-p-Tolyl-guanidine

  • MDL number MFCD04114637
  • Cas Number : 54015-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/n-i-pi-i-tolyl-guanidine/11904?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/n-i-pi-i-tolyl-guanidine/11904?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart