CH6371384382 Chem Impex International, Inc.

tert-Butyl 3-[4-(2-amino-ethyl)phenyl]propionate

  • MDL number MFCD04114526
  • Cas Number : 120225-79-8
  • Molecular Weight : 249.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form Pale yellow oil
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-tert-i-butyl-3-4-2-amino-ethyl-phenyl-propionate/11929?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-tert-i-butyl-3-4-2-amino-ethyl-phenyl-propionate/11929?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart