CH6371384397 Chem Impex International, Inc.

2-Carbethoxy-5-ethylindole

  • MDL number MFCD00022704
  • Cas Number : 37033-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101-107 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form Off-white to yellow crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-carbethoxy-5-ethylindole/11951?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-carbethoxy-5-ethylindole/11951?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart