CH6371384398 Chem Impex International, Inc.

2-Phenylindole-5-sulfonic acid monosodium salt

  • MDL number MFCD04972100
  • Cas Number : 119205-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Sodium 2-phenylindole-5-sulfonate
Purity 0.95
Storage temp. Store at 0-8
Physical Form White to yellow solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-phenylindole-5-sulfonic-acid-monosodium-salt/11953?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-phenylindole-5-sulfonic-acid-monosodium-salt/11953?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart