CH6371384408 Chem Impex International, Inc.

1-Acetyl-2,3-dihydro-1H-indole-2-carboxylic acid

  • MDL number MFCD02258918
  • Cas Number : 82923-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 193-197 °C
Purity 95
Storage temp. Store at 0-8 °C
Physical Form Brown solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-acetyl-2-3-dihydro-1-i-h-i-indole-2-carboxylic-acid/11964?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-acetyl-2-3-dihydro-1-i-h-i-indole-2-carboxylic-acid/11964?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart